EMAIL THIS PAGE TO A FRIEND
Pharmaceutical Development and Technology
HPLC determination of olanzapine and carbamazepine in their nicotinamide cocrystals and investigation of the dissolution profiles of cocrystal tablet formulations.
PMID 24521464