EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Nutrient budgets and effluent characteristics in giant freshwater prawn (Macrobrachium rosenbergii) culture ponds.
PMID 24522829
Abstract
It is important to understand nutrient budgets of aquaculture practices for efficiency of input resources and to utilize all output nutrient sources. The aim of the present study was to develop a nutrient budget for giant freshwater prawn (Macrobrachium rosenbergii) culture ponds. The study was conducted in farmer's ponds (0.25-0.5 ha) of Odisha, India, and the results showed that feed accounted 97% total nitrogen (N), 98.7% total phosphorus (P) and 90% total organic carbon (OC), respectively. The harvested prawn accounted for recovery of 37% N, 10% P and 15% OC, respectively. The N, P and OC accumulated in sediment were 52%, 76%, and 65%, respectively. Nutrient loads in the effluents were 2.22 ± 0.66 kg inorganic N, 0.40 ± 0.15 kg P, and 21.01 ± 6.4 kg OC per ton of prawn production. The present study implicated that high nutrient values observed in both water and sediment provide important opportunities for nutrient reuse through pond sediment applications to croplands as an organic manure, as well as pond water irrigation to crops as a "liquid fertilizer".