EMAIL THIS PAGE TO A FRIEND
The Journal of Neuroscience
Deprivation-induced strengthening of presynaptic and postsynaptic inhibitory transmission in layer 4 of visual cortex during the critical period.
PMID 24523547