EMAIL THIS PAGE TO A FRIEND
Environmental monitoring and assessment
Investigation on diazinon and oxydemeton-methyl residues in cucumbers grown in Kerman greenhouses.
PMID 24526616
Abstract
Pesticides are considered as the most polluting substances. The residue of pesticides in agricultural crops, especially in greenhouse harvests, has been reported critical. Cucumber, considered as a vegetable, is an agricultural product which is commonly found in the Iranian food basket. The current study aims to assess the level of diazinon and oxydemeton-methyl existing in cucumbers sampled from Kerman greenhouses. The pesticide residue was extracted by acetone and dichloromethane. The extracts were cleaned up according to the solid-phase extraction method. The pesticide residues were then determined by capillary gas chromatography with nitrogen-phosphorus detection. The recoveries were 83 and 85 %, the limit of detection was 0.028 and 0.034 mg/kg, and the limits of qualification were 0.093 and 0.113 mg/kg for diazinon and oxydemeton-methyl in cucumber samples, respectively. The median of diazinon residue was detected 0.582 mg/kg, which was 11.64 times the national maximum residue limit (MRL) (0.05 mg/kg), and the median for oxydemeton-methyl was 1.910 mg/kg, being 1.91 times the MRL (1 mg/kg).