EMAIL THIS PAGE TO A FRIEND
The Journal of family practice
PURLs: treating migraine: the case for aspirin.
PMID 24527476
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1440808 Metoclopramide hydrochloride, United States Pharmacopeia (USP) Reference Standard
C14H22ClN3O2 · HCl · H2O