EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Hepatic antioxidant enzymes SOD and CAT of Nile tilapia (Oreochromis niloticus) in response to pesticide methomyl and recovery pattern.
PMID 24531323
Abstract
Hepatic antioxidant enzymes superoxide dismutase (SOD) and catalase (CAT) of Nile tilapia in response to pesticide methomyl and recovery pattern were researched by exposing tilapia to sub-lethal methomyl concentrations of 0, 0.2, 2, 20 and 200 μg/L for 30 days, and then transferred to methomyl-free water for 18 days. Hepatic SOD and CAT were measured at 10 min (day 0), 6, 12, 18, 24 and 30 days after starting the experiment and at 18 days after transferring to methomyl-free water. The results showed hepatic SOD and CAT activities in 2, 20 and 200 μg/L groups were affected significantly, however, that in 0.2 μg/L group didn't change significantly compared to control during 30-day exposure period. Thus it would appear the 0.2 μg/L methomyl might be considered the no observed adverse effect level. Recovery data showed that, for SOD, the effects produced by lower concentration of methomyl 2 μg/L were reversible but not at concentrations higher than 20 μg/L, however, for CAT, the effects produced by all the concentrations were reversible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36159
Methomyl, PESTANAL®, analytical standard
C5H10N2O2S