EMAIL THIS PAGE TO A FRIEND
European journal of nutrition
Effects of regular consumption of vitamin C-rich or polyphenol-rich apple juice on cardiometabolic markers in healthy adults: a randomized crossover trial.
PMID 24531755
Abstract
The aim of the present study was to investigate the effects of the consumption of two cloudy apple juices with different polyphenol and vitamin C contents on antioxidant status, cardiometabolic and inflammation markers in healthy young adults. Twenty subjects, aged 21-29 years, completed a randomized crossover study. At each 4-week intervention period, the volunteers randomly consumed two glasses (2 × 250 mL/day) of either a vitamin C-rich apple juice (VCR) (60 mg/L vitamin C and 510 mg catechin equivalent/L) or a polyphenol-rich (PR) juice (22 mg/L vitamin C and 993 mg catechin equivalent/L). Blood and urine samples were collected throughout the study, and markers of antioxidant status, glucose metabolism, lipid profile and inflammation were measured. The comparison of the post-intervention minus pre-intervention change revealed differential results in HOMA index, total cholesterol, ICAM-1 and VCAM-1 (P < 0.05) across juices. During the VCR period, plasma antioxidant activity (FRAP) increased (P = 0.031), while ICAM-1 and total cholesterol showed a trend to decrease (P = 0.060 and P = 0.094, respectively). During the PR period, plasma insulin and HOMA increased, and total glutathione decreased (P < 0.05). A joint consumption of apple juice natural antioxidants such as vitamin C and polyphenols might provide mild favorable effects on cardiometabolic markers, as compared to apple polyphenols alone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1753
(−)-Epicatechin, ≥90% (HPLC)
C15H14O6
E4018
(−)-Epicatechin, ≥98% (HPLC), from green tea
C15H14O6
68097
(−)-Epicatechin, analytical standard
C15H14O6
D4506
4-(Dimethylamino)cinnamaldehyde, chromogenic reagent for indoles and flavanols
C11H13NO
42251
Apigenin, analytical standard
C15H10O5
01760595
Apigenin, primary reference standard
C15H10O5
32319
Diethylenetriaminepentaacetic acid, for complexometry, ≥99.0%
C14H23N3O10
03940590
Epicatechin, primary reference standard
C15H14O6
P7912
Phloretin, ≥99%
C15H14O5
91897
Phloretin, analytical standard
C15H14O5
79330
Phloroglucinol, ≥99.0% (HPLC)
C6H6O3
P3502
Phloroglucinol, Used to detect the presence of wood fiber.
C6H6O3
P1178
Phloroglucinol, BioReagent, suitable for plant cell culture
C6H6O3
Y0000493
Phloroglucinol (anhydrous), European Pharmacopoeia (EP) Reference Standard
C6H6O3
00200595
Quercetin dihydrate, primary reference standard
C15H10O7 · 2H2O
Y0001009
Quercetin dihydrate, European Pharmacopoeia (EP) Reference Standard
C15H10O7 · 2H2O
V8388
Vincristine sulfate, meets USP testing specifications
C46H56N4O10 · H2SO4
V0400000
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
C46H56N4O10 · H2SO4
V8879
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
C46H56N4O10 · H2SO4