EMAIL THIS PAGE TO A FRIEND
Xenobiotica
Effect of commonly used organic solvents on aldehyde oxidase-mediated vanillin, phthalazine and methotrexate oxidation in human, rat and mouse liver subcellular fractions.
PMID 24533630
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1000000
Methotrexate, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
PHR1396
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
C20H22N8O5
Y0000602
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
Y0001668
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
C20H22N8O5
77634
Mettler-Toledo Calibration substance ME 51143093, Vanillin, traceable to primary standards (LGC)
C8H8O3
P38706
Phthalazine, 98%
C8H6N2
P39001
Phthalazone, 99%
C8H6N2O
94770
Vanillic acid, purum, ≥97.0% (HPLC)
C8H8O4
H36001
Vanillic acid, 97%
C8H8O4
W398802
Vanillic acid, ≥97%, FG
C8H8O4
68654
Vanillic acid, certified reference material, TraceCERT®
C8H8O4
V2375
Vanillin
C8H8O3
30304
Vanillin, TraceCERT®, certified reference material
C8H8O3
94752
Vanillin, tested according to Ph.Eur.
C8H8O3
PHR1245
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
V1104
Vanillin, ReagentPlus®, 99%
C8H8O3
W310700
Vanillin, ≥97%, FCC, FG
C8H8O3
W310727
Vanillin, natural, ≥97%, FCC, FG
C8H8O3
V0050000
Vanillin, European Pharmacopoeia (EP) Reference Standard
C8H8O3
PHR1091
Vanillin melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H8O3
1711009
Vanillin Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
C8H8O3