EMAIL THIS PAGE TO A FRIEND
Journal of Agricultural and Food Chemistry
Bathochromic and hyperchromic effects of aluminum salt complexation by anthocyanins from edible sources for blue color development.
PMID 24547952