EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
PARP1-dependent recruitment of KDM4D histone demethylase to DNA damage sites promotes double-strand break repair.
PMID 24550317
Abstract
Members of the lysine (K)-specific demethylase 4 (KDM4) A-D family of histone demethylases are dysregulated in several types of cancer. Here, we reveal a previously unrecognized role of KDM4D in the DNA damage response (DDR). We show that the C-terminal region of KDM4D mediates its rapid recruitment to DNA damage sites. Interestingly, this recruitment is independent of the DDR sensor ataxia telangiectasia mutated (ATM), but dependent on poly (ADP-ribose) polymerase 1 (PARP1), which ADP ribosylates KDM4D after damage. We demonstrate that KDM4D is required for efficient phosphorylation of a subset of ATM substrates. We note that KDM4D depletion impairs the DNA damage-induced association of ATM with chromatin, explaining its effect on ATM substrate phosphorylation. Consistent with an upstream role in DDR, KDM4D knockdown disrupts the damage-induced recombinase Rad51 and tumor protein P53 binding protein foci formation. Consequently, the integrity of homology-directed repair and nonhomologous end joining of DNA breaks is impaired in KDM4D-deficient cells. Altogether, our findings implicate KDM4D in DDR, furthering the links between the cancer-relevant networks of epigenetic regulation and genome stability.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0996 PARP-1 human, recombinant, expressed in E. coli