EMAIL THIS PAGE TO A FRIEND
Journal of dietary supplements
Potential oil yield, fatty acid composition, and oxidation stability of the hempseed oil from four Cannabis sativa L. cultivars.
PMID 24552275
Abstract
The cultivation of four industrial hemp cultivars (Felina 32, Chamaeleon, Uso31, and Finola) was investigated for oil production in the north-east of Italy along two years. The oils of all cultivars resulted in rich amount of linoleic acid (ω-6) and α-linolenic acid (ω-3). Felina 32 and Chamaeleon oils exhibited the highest amount of linoleic acid (59%) and α-linolenic acid (18%). Finola and Uso31 oils resulted in the richest of γ-linolenic acid (5-6%). All hempseed oils presented high oxidation stability and an acceptable initial quality. It is suggested that these oils can be used to produce EFA dietary supplements high in ω-6 and ω-3 of vegetal origin.