EMAIL THIS PAGE TO A FRIEND
Cerebral cortex (New York, N.Y. : 1991)
GluN2A-/- Mice Lack Bidirectional Synaptic Plasticity in the Dentate Gyrus and Perform Poorly on Spatial Pattern Separation Tasks.
PMID 24554729
Abstract
The different secondary subunits of the N-methyl-d-aspartate (NMDA) receptor each convey unique biophysical properties to the receptor complex, and may be key in determining the functional role played by NMDA receptors. In the hippocampus, the GluN2A and GluN2B subunits are particularly abundant; however, their exact roles in synaptic plasticity and behavior remain controversial. Here, we show that mice carrying a deletion for the GluN2A subunit (GluN2A(-/-)) demonstrate a severely compromised NMDA to AMPA receptor current ratio in granule cells from the dentate gyrus (DG), while granule cell morphology is unaltered. This deficit is accompanied by significant impairments in both LTP and LTD in the DG, whereas only minor impairments are observed in the CA1. In accordance with these hippocampal region-specific deficits, GluN2A(-/-) mice show impaired performance on the DG-associated task of spatial pattern separation. In contrast, GluN2A(-/-) mice show no deficit in temporal pattern separation, a process associated with CA1 functioning. Thus, our results establish the GluN2A subunit as a significant contributor to both bidirectional synaptic plasticity and spatial pattern separation in the DG.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14340
(+)-Bicuculline, ≥97.0% (TLC)
C20H17NO6
A5282
DL-2-Amino-5-phosphonopentanoic acid, solid
C5H12NO5P