EMAIL THIS PAGE TO A FRIEND
Molecular Biology of the Cell
Kdm3a lysine demethylase is an Hsp90 client required for cytoskeletal rearrangements during spermatogenesis.
PMID 24554764
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0067
IOX1, ≥97% (HPLC)
C10H7NO3