EMAIL THIS PAGE TO A FRIEND
Microvascular research
Thrombospondin-1 domain-containing peptide properdistatin improves vascular function in human melanoma xenografts.
PMID 24555949
Abstract
Properdistatin is a novel peptide derived from the thrombospondin-1 domain of the plasma protein properdin. The purpose of this study was to investigate the effect of properdistatin treatment on the morphology and function of tumor vasculature. A-07 human melanoma xenografts grown in dorsal window chambers were used as preclinical model. Tumors were treated with 80 mg/kg/day properdistatin or vehicle for 4 days. Morphologic parameters of tumor vascular networks were assessed from high-resolution transillumination images, and tumor blood supply time and plasma velocities were assessed from first-pass imaging movies recorded after a bolus of 155 kDa tetramethylrhodamine isothiocyanate-labeled dextran had been administered intravenously. Properdistatin-treated tumors showed reduced density of small-diameter vessels, reduced blood supply time, and increased plasma velocities. In conclusion, properdistatin treatment inhibited angiogenesis and improved vascular function in A-07 tumors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3886
Fentanyl citrate salt
C22H28N2O · C6H8O7