EMAIL THIS PAGE TO A FRIEND
Cancer treatment reviews
nab-Paclitaxel in combination with biologically targeted agents for early and metastatic breast cancer.
PMID 24560997
Abstract
Taxanes are highly active chemotherapeutic agents used in the treatment of early-stage and metastatic breast cancer. Novel formulations have been developed to improve efficacy and decrease toxicity associated with these cytotoxic agents. nab-Paclitaxel is a biologically interactive, solvent-free, 130-nm-sized albumin-bound paclitaxel, developed to avoid the Cremophor vehicle used in solvent-based paclitaxel. Based on a pivotal phase 3 study, nab-paclitaxel was shown to be safely infused at a significantly higher dose of paclitaxel than the doses used with standard paclitaxel therapy, and had a shorter infusion time, no premedication, and higher response rates. It is now approved in the United States for treatment of breast cancer after failure of combination chemotherapy for metastatic disease or relapse within 6 months of adjuvant therapy, and has demonstrated promising efficacy and favorable tolerability. Recently, several phase 2 and 3 studies have suggested a role for nab-paclitaxel in combination with biologically targeted agents for the treatment of early- and late-stage breast cancer. This review will discuss the findings of clinical trials evaluating nab-paclitaxel in combination with biologically targeted therapeutic agents for breast cancer in the neoadjuvant, adjuvant, and metastatic settings.