EMAIL THIS PAGE TO A FRIEND
Nature cell biology
Nascent chromatin capture proteomics determines chromatin dynamics during DNA replication and identifies unknown fork components.
PMID 24561620
Abstract
To maintain genome function and stability, DNA sequence and its organization into chromatin must be duplicated during cell division. Understanding how entire chromosomes are copied remains a major challenge. Here, we use nascent chromatin capture (NCC) to profile chromatin proteome dynamics during replication in human cells. NCC relies on biotin-dUTP labelling of replicating DNA, affinity purification and quantitative proteomics. Comparing nascent chromatin with mature post-replicative chromatin, we provide association dynamics for 3,995 proteins. The replication machinery and 485 chromatin factors such as CAF-1, DNMT1 and SUV39h1 are enriched in nascent chromatin, whereas 170 factors including histone H1, DNMT3, MBD1-3 and PRC1 show delayed association. This correlates with H4K5K12diAc removal and H3K9me1 accumulation, whereas H3K27me3 and H3K9me3 remain unchanged. Finally, we combine NCC enrichment with experimentally derived chromatin probabilities to predict a function in nascent chromatin for 93 uncharacterized proteins, and identify FAM111A as a replication factor required for PCNA loading. Together, this provides an extensive resource to understand genome and epigenome maintenance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-595
Anti-acetyl-Histone H4 (Lys12) Antibody, Use Anti-acetyl-Histone H4 (Lys12) Antibody (Rabbit Polyclonal Antibody) validated in ChIP, DB, WB, ChIP-seq to detect acetyl-Histone H4 (Lys12) also known as H4K12Ac, Histone H4 (acetyl K12).
05-858
Anti-Histone H4 Antibody, pan, clone 62-141-13, rabbit monoclonal, Anti-Histone H4 Antibody, pan, clone 62-141-13 is a Rabbit Monoclonal Antibody for detection of Histone H4 also known as Histone H4 & has been validated in ChIP, WB, Mplex, ChIP-seq.
07-448
Anti-monomethyl-Histone H3 (Lys27) Antibody, Anti-monomethyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody to detect Histone H3 monomethylated at lysine 27. This purified Ab, also known as Anti-H3K27me1 is specificity verified by dot blot (DB), published in peer reviewed journals and validated in ICC, WB, ChIP, ChIP-seq.
07-450
Anti-monomethyl-Histone H3 (Lys9) Antibody, Anti-monomethyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 monomethylated on lysine 9 (H3K9me1). This purified antibody is specificity verified by dot blot (DB), published in peer reviewed journals, and has been validated in ICC, PIA & WB.
07-442
Anti-trimethyl-Histone H3 (Lys9) Antibody, Use Anti-trimethyl-Histone H3 (Lys9) Antibody (rabbit polyclonal antibody) validated in DB, ICC, Mplex, PIA, WB, ChIP-seq to detect trimethyl-Histone H3 (Lys9) also known as H3K9me3, Histone H3 (tri methyl K9).
HPA040176
Anti-FAM111A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution