EMAIL THIS PAGE TO A FRIEND
Endocrinology
Metabolic influences on reproduction: adiponectin attenuates GnRH neuronal activity in female mice.
PMID 24564393
Abstract
Metabolic dysfunctions are often linked to reproductive abnormalities. Adiponectin (ADP), a peripheral hormone secreted by white adipose tissue, is important in energy homeostasis and appetite regulation. GnRH neurons are integral components of the reproductive axis, controlling synthesis, and release of gonadotropins. This report examined whether ADP can directly act on GnRH neurons. Double-label immunofluorescence on brain sections from adult female revealed that a subpopulation of GnRH neurons express ADP receptor (AdipoR)2. GnRH/AdipoR2+ cells were distributed throughout the forebrain. To determine the influence of ADP on GnRH neuronal activity and the signal transduction pathway of AdipoR2, GnRH neurons maintained in explants were assayed using whole-cell patch clamping and calcium imaging. This mouse model system circumvents the dispersed distribution of GnRH neurons within the forebrain, making analysis of large numbers of GnRH cells possible. Single-cell PCR analysis and immunocytochemistry confirmed the presence of AdipoR2 in GnRH neurons in explants. Functional analysis revealed 20% of the total GnRH population responded to ADP, exhibiting hyperpolarization or decreased calcium oscillations. Perturbation studies revealed that ADP activates AMP kinase via the protein kinase Cζ/liver kinase B1 pathway. The modulation of GnRH neuronal activity by ADP demonstrated in this report directly links energy balance to neurons controlling reproduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
05164
(Aminomethyl)phosphonic acid, analytical standard
CH6NO3P
17932
1,3-Butadiene, puriss., ≥99.5% (GC)
C4H6
295035
1,3-Butadiene, ≥99%
C4H6
695580
1,3-Butadiene solution, 20 wt. % in toluene
C4H6
695904
1,3-Butadiene solution, 15 wt. % in hexane
C4H6
CDS020502
2-Aminoimidazole, AldrichCPR
C3H5N3
HPA057769
Anti-CD1E antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA028900
Anti-CD82 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PRS4073
Anti-KAI1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
GW22152A Anti-RRM2 antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB2701764
Anti-rrm2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA056994
Anti-RRM2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411214
Anti-RRM2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
A5835
γ-Aminobutyric acid, BioXtra, ≥99%
C4H9NO2
A2129
γ-Aminobutyric acid, ≥99%
C4H9NO2
03835
γ-Aminobutyric acid, analytical standard
C4H9NO2
PHR1351
Inositol, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
WH0003732M1
Monoclonal Anti-CD82 antibody produced in mouse, clone 4F2, purified immunoglobulin, buffered aqueous solution
SAB4700755 Monoclonal Anti-CD82 antibody produced in mouse, clone C33, purified immunoglobulin, buffered aqueous solution
WH0006241M1
Monoclonal Anti-RRM2 antibody produced in mouse, clone 1E1, purified immunoglobulin, buffered aqueous solution
SAB1404339
Monoclonal Anti-RRM2, (N-terminal) antibody produced in mouse, clone 1E1, ascites fluid
Y0001404
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
C4H9NO2