EMAIL THIS PAGE TO A FRIEND
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology
Is the consistency more important than the ingredients for steroid treatment in eosinophilic esophagitis?
PMID 24566294