EMAIL THIS PAGE TO A FRIEND
JAMA pediatrics
Antenatal acetaminophen use and attention-deficit/hyperactivity disorder: an interesting observed association but too early to infer causality.
PMID 24566519