EMAIL THIS PAGE TO A FRIEND
The American journal of tropical medicine and hygiene
Clinical and immunological outcome in cutaneous leishmaniasis patients treated with pentoxifylline.
PMID 24567316
Abstract
Pentoxifylline is a tumor necrosis factor-α (TNF-α) inhibitor that also attenuates the immune response and decreases tissue inflammation. The association of pentoxifylline with antimony improves the cure rate of mucosal and cutaneous leishmaniasis. In this randomized and double blind pilot trial, cure rate was higher, although not significant, in patients who received antimony plus pentoxifylline than in those patients receiving antimony plus placebo. A significant decrease in TNF-α and interferon-γ (IFN-γ) levels during therapy was more pronounced in the antimony plus pentoxifylline group, whereas CCL-3 (Chemokine [C-C motif] ligand 3) decreased similarly in both groups. The increased levels of CXCL-9 (Chemokine [C-X-C motif] ligand 9) during therapy were lower in the antimony plus pentoxifylline group. Therapy with pentoxifylline modifies cytokines and chemokines production, which may be associated with therapeutic outcome.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF17616214
Antimony, rod, 6mm, diameter 6mm, 99.999%
Sb
GF76817616
Antimony, foil, 50x50mm, thickness 2mm, hot-pressed
Sb
GF15127961
Antimony, foil, light tested, 25x25mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF34540185
Antimony, foil, light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF72464834
Antimony, foil, not light tested, 100x100mm, thickness 0.015mm, permanent polyester support, 95+%
Sb
GF97030403
Antimony, foil, not light tested, 100x100mm, thickness 0.01mm, permanent polyester support, 95+%
Sb
GF90469884
Antimony, foil, not light tested, 100x100mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF45762789
Antimony, foil, not light tested, 100x100mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF89164873
Antimony, foil, not light tested, 25x25mm, thickness 0.002mm, permanent polyester support, 95+%
Sb
GF16935396
Antimony, foil, not light tested, 25x25mm, thickness 0.005mm, permanent polyester support, 95+%
Sb
GF97807427
Antimony, foil, not light tested, 25x25mm, thickness 0.015mm, permanent polyester support, 95+%
Sb
GF83929674
Antimony, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF04779894
Antimony, foil, not light tested, 50x50mm, thickness 0.002mm, permanent polyester support, 95+%
Sb
GF07563203
Antimony, foil, not light tested, 50x50mm, thickness 0.005mm, permanent polyester support, 95+%
Sb
GF66083362
Antimony, foil, not light tested, 50x50mm, thickness 0.015mm, permanent polyester support, 95+%
Sb
GF72466803
Antimony, foil, not light tested, 50x50mm, thickness 0.025mm, permanent polyester support, 95+%
Sb
GF16928310
Antimony, foil, not light tested, 50x50mm, thickness 0.05mm, permanent polyester support, 95+%
Sb
GF87764013
Antimony, microfoil, 25x25mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Sb
GF70501788
Antimony, microfoil, 50x50mm, thinness 0.5μm, specific density 334.5μg/cm2, 6 micron aluminum permanent support, 95+%
Sb
GF59022592
Antimony, rod, 15mm, diameter 2.0mm, 99.99%
Sb
GF96258437
Antimony, rod, 20mm, diameter 4.7mm, 99.999%
Sb
GF20324865
Antimony, rod, 25mm, diameter 2.0mm, 99.99%
Sb
GF87920343
Antimony, rod, 50mm, diameter 2.0mm, 99.99%
Sb
GF01771118
Antimony, rod, 50mm, diameter 4.7mm, 99.999%
Sb
GF83770094
Antimony, foil, not light tested, 25x25mm, thickness 0.01mm, permanent polyester support, 95+%
Sb
GF46574220
Antimony, foil, not light tested, 50x50mm, thickness 0.01mm, permanent polyester support, 95+%
Sb