EMAIL THIS PAGE TO A FRIEND
Toxicology mechanisms and methods
3-Aminobenzamide--a PARP inhibitor enhances the sensitivity of peripheral blood micronucleus and comet assays in mice.
PMID 24568202
Abstract
DNA repair is an essential outcome of DNA damage, which may compromise the end point of various in vitro and in vivo test systems of the genotoxicity evaluation. poly(ADP-ribose) polymerase (PARP) enzymes have an essential role in DNA repair. Here, we investigated the effect of 3-AB, a PARP inhibitor on the sensitivity of comet and PBMN assays. This study aimed to enhance the sensitivity of the comet and peripheral blood micronucleus (PBMN) assays using 3-aminobenzamide (3-AB), a well-characterized PARP inhibitor. Cyclophosphamide (CP, 50 mg/kg), 5-flourouracil (5-FU, 25 mg/kg), zidovudine (AZT, 400 mg/kg) and furosemide (FUR, 60 mg/kg) were selected as genotoxins. 3-AB was given every 8 h with the first dose given 2 h before the genotoxin treatment. For the PBMN assay, small amount of blood was taken from the tail tip of each animal and smears were prepared. The comet assay was performed in PBL, bone marrow and liver. In the comet as well as PBMN assay, 3-AB pre-treatment enhanced the extent of DNA damage in all the combination groups (3-AB + CP, 3-AB + 5-FU and 3-AB + AZT) compared to CP, 5-FU and AZT per se. 3-AB also enhanced the DNA damage caused by FUR in the bone marrow and liver. This study results clearly demonstrate that the pretreatment with 3-AB (30 mg/kg) significantly enhances the sensitivity of the PBMN and comet assays. This model may be useful for the detection of marginally active DNA damaging agents.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0788
3-Aminobenzamide, ≥99% (TLC)
C7H8N2O