EMAIL THIS PAGE TO A FRIEND
Tissue engineering. Part A
rhPDGF-BB via ERK pathway osteogenesis and adipogenesis balancing in ADSCs for critical-sized calvarial defect repair.
PMID 24568547
Abstract
Adipose-derived stem cells (ADSCs) with the capacity of differentiating into osteo-like cells have been widely investigated for bone tissue engineering as a novel seed cell source. Recombinant human platelet-derived growth factor (rhPDGF-BB) is a clinically proven growth factor with the potential of promoting cell proliferation and osteogenic differentiation during the bone regeneration process. In this study, we investigated the effects of rhPDGF-BB on the proliferation and osteogenic and adipogenic differentiation of rat ADSCs and explored whether the extracellular signal-related kinase (ERK) signaling pathway might be involved. We found that rhPDGF-BB significantly enhanced ADSCs proliferation and osteogenic differentiation, as detected by MTT, real-time polymerase chain reaction (PCR), ALP activity assays, and calcium deposition in vitro, in concert with ERK pathway activation. In contrast, the adipogenesis of ADSCs, as detected by real-time PCR and Oil Red O staining, was suppressed in the presence of rhPDGF-BB. Furthermore, with the supplement of the ERK inhibitor PD98059, cell proliferation and osteogenesis were reduced; as expected, adipogenesis was enhanced. Subsequently, for the first time, we evaluated the effect of ADSCs associated with rhPDGF-BB on bone regeneration in a critical-sized rat calvarial defect model with silk scaffold as a carrier. Micro-computed tomography and histological analyses exhibited dramatically more new bone formation and trabecular number in the Silk/PDGF/ADSC group. These data indicated that rhPDGF-BB promoted cell proliferation and osteogenic differentiation and suppressed adipogenic differentiation in vitro via ERK pathway and that ADSCs associated with rhPDGF-BB could be a promising tissue-engineered construct for craniofacial bone regeneration in vivo.