EMAIL THIS PAGE TO A FRIEND
International journal of clinical pharmacology and therapeutics
Effect of dual bronchodilation with QVA149 on cardiac safety in healthy volunteers.
PMID 24569129
Abstract
QVA149 is a dual bronchodilator, containing a fixed-dose combination of the long-acting β2-agonist indacaterol and long-acting muscarinic antagonist glycopyrronium, for the treatment of chronic obstructive pulmonary disease (COPD). Here we assess the potential of QVA149 (440/200 μg) at 4-fold the therapeutic dose for causing cardiac pharmacodynamic (PD) effects. This double-blind, randomized study estimated the time-matched largest heart rate (HR) change and average HR change (over 24 hours) from baseline for QVA149 vs. placebo in healthy subjects. Similar analyses were done for QVA149 vs. indacaterol 600 μg, glycopyrronium 200 μg, and salmeterol 200 μg. The time-matched and average change from baseline in QT interval corrected for HR using Fridericia's formula (QTcF), effects on serum potassium and blood glucose, pharmacokinetic (PK) parameters, and safety were also assessed. Of 50 subjects randomized, 43 completed the study. QVA149, when compared with placebo, showed the time-matched largest mean increase and decrease in HR of 5.69 bpm and -2.51 bpm, respectively, and average HR change from baseline of 0.62 bpm. QVA149 showed no tachycardic potential compared with indacaterol and no relevant tachycardic effect compared with glycopyrronium. No consistent differences were seen in the time-matched largest mean change and average change from baseline in QTcF for QVA149 vs. other treatments. There were no relevant effects of QVA149 on serum potassium and blood glucose. There was no apparent PK/PD relationship between the observed exposures to indacaterol and glycopyrronium in QVA149 on HR and QTcF. There were no deaths or serious adverse events. Overall, short-term administration of QVA149 showed a good cardiovascular safety and tolerability profile in healthy subjects.