EMAIL THIS PAGE TO A FRIEND
Journal of helminthology
Rohdella amazonica n. sp. (Aspidogastrea: Aspidogastridae) from the Amazoninan banded puffer fish Colomesus psittacus (Bloch & Schneider, 1801).
PMID 24572176
Abstract
Aspidogastreans are commonly found infecting freshwater and marine molluscs, teleosts fishes and freshwater turtles. The subclass comprises four families - Rugogastridae Schell 1973, Stichocotylidae Faust & Tang 1936, Multicalycidae Gibson & Chinabut 1984 and Aspidogastridae Poche 1907 - and it is characterized by the presence of a ventral adhesive disc divided into rows of alveoli. In the current work, using light and scanning electron microscopy and molecular approaches, a new species of Aspidogastridae of the genus Rohdella Gibson & Chinabut, 1984, is described as a parasite of Colomesus psittacus in Brazil. The new taxon is distinguishable by the presence of oesophageal glands, teguments covered by ciliated papillae, and the position and shape of the hermaphroditic duct. The present work describes the third species of the genus Rohdella, thereby adding new morphological and molecular data regarding Aspidogastridae.