EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Histomorphologic and histomorphometric characteristics of zygapophyseal joint remodeling in ankylosing spondylitis.
PMID 24574301
Abstract
To unravel the mechanisms that control bony ankylosis in ankylosing spondylitis (AS). Histomorphologic and histomorphometric analyses were performed on zygapophyseal joints obtained from 18 patients with AS, 9 patients with osteoarthritis (OA), and 10 cadaver donors without a rheumatic disease (controls). The proteoglycan content of the cartilage was determined by Safranin O staining and the chondrocyte apoptosis according to caspase 3 expression. AS joints were categorized into 3 groups according to the morphology of the joint surfaces and joint space: group 1 were joints with an open joint space, group 2 were joints with cartilaginous fusion, and group 3 were joints with bony fusion of the joint surfaces. Progressive loss of the joint space from group 1 joints to group 3 joints suggests that this grouping corresponds to sequential stages of joint remodeling. Cartilage thickness and subchondral bone plate thickness declined from group 1 to group 3 (P < 0.01). Increased chondrocyte apoptosis rates were found in groups 1 and 2 (P < 0.05), while in group 3, a reduction in the proteoglycan content was found (P < 0.001). Bone marrow replacement and invasion of the subchondral bone plate by fibrous tissue was found predominantly in AS joints in group 2. Cartilage degeneration, indicated by cartilage thinning, enhanced chondrocyte apoptosis, and proteoglycan loss, and subchondral bone thinning, promoted by invasion of the subchondral bone plate by a fibrous tissue originating from the bone marrow, are hallmarks of joint remodeling in AS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8482
Anti-ACP5 (132-145) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB1401002
Anti-ACP5 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1401003
Anti-ACP5 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501669
Anti-CASP3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA002643
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV00021
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
SAB4503291
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
AV41929
Anti-CD40 antibody produced in rabbit, affinity isolated antibody
HPA031567
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA031568
Anti-CD40 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
C6487 Anti-CD40 Ligand antibody produced in goat, affinity isolated antibody, lyophilized powder
SAB2103449
Anti-CD40, (N-terminal) antibody produced in rabbit, affinity isolated antibody
SAB2100377
Anti-CD40LG antibody produced in rabbit, affinity isolated antibody
HPA045827
Anti-CD40LG antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700234
Anti-Ncam1 (N-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2501672
Anti-NCAM1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4300660
Anti-NCAM1 antibody produced in rabbit, affinity isolated antibody
HPA015015
Anti-SCYL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701096
Anti-SCYL1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA024529
Anti-TDRD7 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501057
Anti-TRAF2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
HPA009972
Anti-TRAF2 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA010634
Anti-TRAF2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500375
Anti-TRAF2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1402681
Monoclonal Anti-ACP5 antibody produced in mouse, clone 2D9, purified immunoglobulin, buffered aqueous solution
WH0000836M2
Monoclonal Anti-CASP3 antibody produced in mouse, clone 4D3, purified immunoglobulin, buffered aqueous solution
C5737 Monoclonal Anti-Caspase 3 antibody produced in mouse, clone 84803.111, purified immunoglobulin, lyophilized powder
WH0000958M1
Monoclonal Anti-CD40 antibody produced in mouse, clone 1G1, purified immunoglobulin, buffered aqueous solution
AMAB90905
Monoclonal Anti-CD40 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1673, purified immunoglobulin, buffered aqueous glycerol solution
WH0000959M1
Monoclonal Anti-CD40LG antibody produced in mouse, clone 2E2, purified immunoglobulin, buffered aqueous solution