EMAIL THIS PAGE TO A FRIEND
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego
[Diabetes mellitus-related oxidative stress and its parameters in saliva].
PMID 24575653
Abstract
The uncontrolled accumulation of glucose and fatty acids in the human body without the possibility of any cells utilizing them as metabolic substrates is a typical occurrence in the process of diabetes mellitus. High concentration of mentioned substances leads to oxidative stress and that reasons in numerous complications as a result of diabetes which are often demonstrated in the oral cavity as periodontal inflammations. There are many ways to study the biomarkers of oxidative stress such as: superoxide dismutase (SOD), catalase, glutathione peroxidase, glutathione reductase, glutathione, vitamins, lipid peroxidation, concentration of nitrites, non-enzymatic glycosylation of proteins and hyperglycaemia in diabetes. Considering the attainability and simplicity required to collect, saliva is a popular research material used to test these biomarkers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3515 Catalase from Aspergillus niger, ammonium sulfate suspension, ≥4,000 units/mg protein
C1345 Catalase from bovine liver, powder, suitable for cell culture, 2,000-5,000 units/mg protein
C9322 Catalase from bovine liver, lyophilized powder, 2,000-5,000 units/mg protein
C30 Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein
C40 Catalase from bovine liver, lyophilized powder, ≥10,000 units/mg protein
C3155 Catalase from bovine liver, aqueous solution, ≥30,000 units/mg protein
C100 Catalase from bovine liver, aqueous suspension, 40,000-60,000 units/mg protein (E1%/405)
E3289 Catalase from bovine liver, ≥20000 units/mg protein, lyophilized powder
C3556 Catalase from human erythrocytes, ≥90% (SDS-PAGE), buffered aqueous solution, ≥30,000 units/mg protein
C9284 Catalase−Agarose, pH 5.4(isoelectric point), ammonium sulfate suspension, enzyme from bovine liver
G6137 Glutathione Peroxidase from bovine erythrocytes, lyophilized powder, ≥300 units/mg protein
G4013 Glutathione Peroxidase from human erythrocytes, lyophilized powder, ≥30 units/mg protein
S9697
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
S5395
Superoxide Dismutase from bovine erythrocytes, BioReagent, ≥3,000 units/mg protein, suitable for cell culture, lyophilized powder
S7571
Superoxide Dismutase from bovine erythrocytes, lyophilized powder, ≥3,000 units/mg protein
S7446
Superoxide Dismutase from bovine erythrocytes, BioUltra, lyophilized powder, ≥4,500 units/mg protein, ≥97% (SDS-PAGE)
S8160
Superoxide Dismutase from bovine liver, lyophilized powder, ≥1500 units/mg protein
S8409
Superoxide Dismutase from bovine liver, ammonium sulfate suspension, 2,000-6,000 units/mg protein (biuret)
S5639
Superoxide Dismutase from Escherichia coli, lyophilized powder, ≥1,000 units/mg protein
S9636
Superoxide Dismutase from human erythrocytes, essentially salt-free, lyophilized powder, ≥2,500 units/mg protein
S9076 Superoxide Dismutase I human, recombinant, expressed in E. coli