EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
Cartilage tissue engineering application of injectable gelatin hydrogel with in situ visible-light-activated gelation capability in both air and aqueous solution.
PMID 24575844