EMAIL THIS PAGE TO A FRIEND
Tissue Engineering: Part A
A tissue engineering approach for periodontal regeneration based on a biodegradable double-layer scaffold and adipose-derived stem cells.
PMID 24575867