EMAIL THIS PAGE TO A FRIEND
Arthritis & rheumatology (Hoboken, N.J.)
Inhibition of spermidine/spermine N1-acetyltransferase activity: a new therapeutic concept in rheumatoid arthritis.
PMID 24578214
Abstract
Changes in polyamine-modulated factor 1 (PMF-1) promoter methylation might favor the expression of spermidine/spermine N1-acetyltransferase 1 (SSAT-1), causing excessive consumption of S-adenosyl methionine (SAM). This study was undertaken to evaluate the effect of SSAT-1 activity inhibition, either alone or in combination with SAM. Synovial fibroblasts were isolated from patients with rheumatoid arthritis (RA) or osteoarthritis (OA). PMF-1 promoter methylation was determined by pyrosequencing. Small interfering RNAs (siRNAs) against SSAT-1 were transfected weekly in RA synovial fibroblasts (RASFs). In addition, synovial fibroblasts were treated with diminazene aceturate (DA), an inhibitor of SSAT-1. SSAT-1, 5-methylcytosine (5-MeC), adenosyl methionine decarboxylase (AMD), PMF-1, DNA methyltransferase 1 (DNMT-1), CXCL12, β1 integrin, and CD44 levels were measured by flow cytometry. Putrescine levels were determined by colorimetry. Levels of matrix metalloproteinases were measured by enzyme-linked immunosorbent assay. Cell adhesion was tested. The SCID mouse model of RA was used to monitor the invasiveness of RASFs. RASFs showed elevated SSAT-1, AMD, and PMF-1 levels. However, PMF-1 promoter methylation was unchanged. Transfection of siRNA targeting SSAT-1 increased 5-MeC levels within 21 days. Similarly, DA increased 5-MeC levels in RASFs. In addition, DA increased the levels of DNMT-1, decreased the levels of AMD, putrescine, activation markers, and MMP-1, and altered the adhesion of RASFs. DA was more efficient in RASFs with higher levels of SSAT-1. Most interestingly, the combination of DA and SAM reduced the invasiveness of RASFs by 70%. The use of DA alone or in combination with SAM/L-methionine might introduce a new therapeutic concept in RA. This is the first therapy that would directly target RASFs and thereby inhibit ongoing joint destruction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7770
Diminazene aceturate, analytical standard
C14H15N7 · 2C4H7NO3