EMAIL THIS PAGE TO A FRIEND
Drug and Chemical Toxicology
The effect of cloudy apple juice on hepatic and mammary gland phase I and II enzymes induced by DMBA in female Sprague-Dawley rats.
PMID 24580095
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
237329
1-Chloro-2,4-dinitrobenzene, ≥99%
C6H3ClN2O4
138630
1-Chloro-2,4-dinitrobenzene, 97%
C6H3ClN2O4
D1878
2,6-Dichloroindophenol sodium salt hydrate, suitable for vitamin C determination, BioReagent
C12H6Cl2NNaO2 · xH2O
M1390
3,3′-Methylene-bis(4-hydroxycoumarin)
C19H12O6
D3254
7,12-Dimethylbenz[a]anthracene, ≥95%
C20H16
AV41405
Anti-CYP1A1 (AB3) antibody produced in rabbit, affinity isolated antibody
SAB2501753
Anti-cyp1a1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1408926
Anti-CYP1A1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1410273
Anti-CYP1A1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB2501712
Anti-Cyp1a2 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB4500579
Anti-Cytochrome P450 1A1 antibody produced in rabbit, affinity isolated antibody
SAB5300119
Monoclonal Anti-CYP1A1 antibody produced in mouse, clone 6G5, purified immunoglobulin, buffered aqueous solution
424455
Resorufin, Dye content 95 %
C12H7NO3
73144
Resorufin, 95%
C12H7NO3
E3763
Resorufin ethyl ether
C14H11NO3
46121
Resorufin ethyl ether, suitable for fluorescence, ≥95% (UV)
C14H11NO3
M1544
Resorufin methyl ether
C13H9NO3
69125
Resorufin methyl ether, suitable for fluorescence, ≥98.0% (HPLC)
C13H9NO3
119814
Sodium 2,6-dichloroindophenolate hydrate, ACS reagent
C12H6Cl2NNaO2 · xH2O