EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Analysis of the conformation and ligand-binding properties of the activation segment of pig procarboxypeptidase A.
PMID 2458099
Abstract
The isolated activation segment of pig procarboxypeptidase A binds two Tb3+ ions in a strong and specific way. In contrast, the binding of Ca2+, Cd2+ and Mg2+ is weak. The binding of Tb3+ increases the resistance of the isolated activation segment against proteolysis and competes for the binding of the carbocyanine dye Stains-All. This dye forms complexes with the activation segment showing spectral properties similar to those observed with EF-hand structures. The presented results support a previous hypothesis on the existence of two regions in the activation segment of pancreatic procarboxypeptidases structurally related to Ca2+-binding domains of the EF-hand protein family.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9379
Stains-All, ~95%
C30H27BrN2S2
230960
Stains-All, Dye content 95 %
C30H27BrN2S2