EMAIL THIS PAGE TO A FRIEND
Vaccine
The assembly conformation of rotavirus VP6 determines its protective efficacy against rotavirus challenge in mice.
PMID 24583002
Abstract
Viral protein assemblies have shown to be superior immunogens used in commercial vaccines. However, little is known about the effect of protein assembly structure in immunogenicity and the protection conferred by a vaccine. In this work, rotavirus VP6, a polymorphic protein that assembles into nanotubes, icosahedra (dlRLP) or trimers was used to compare the immune response elicited by three different assemblies. VP6 is the most antigenic and abundant rotavirus structural protein. It has been demonstrated that antibodies against VP6 interfere with the replication cycle of rotavirus, making it a vaccine candidate. Groups of mice were immunized with either nanotubes, dlRLP or trimers and the humoral response (IgG and IgA titers) was measured. Immunized mice were challenged with EDIM rotavirus and protection against rotavirus infection, measured as viral shedding, was evaluated. Immunization with nanotubes resulted in the highest IgG titers, followed by immunization with dlRLP. While immunization with one dose of nanotubes was sufficient to reduce viral shedding by 70%, two doses of dlRLP or trimers were required to obtain a similar protection. The results show that the type of assembly of VP6 results in different humoral responses and protection efficacies against challenge with live virus. This information is important for the design of recombinant vaccines in general.