EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
High prevalence of mTOR complex activity can be targeted using Torin2 in papillary thyroid carcinoma.
PMID 24583924
Abstract
The mammalian target of rapamycin (mTOR) signaling cascade is a key regulatory pathway controlling initiation of messenger RNA in mammalian cells. Although dysregulation of mTOR signaling has been reported earlier in cancers, there is paucity of data about mTOR expression in papillary thyroid carcinoma (PTC). Therefore, in this study, we investigated the presence of mTORC2 and mTORC1 complexes in a large cohort of >500 PTC samples. Our clinical data showed the presence of active mTORC1 and mTORC2 in 81 and 39% of PTC samples, respectively. Interestingly, coexpression of mTORC1 and mTORC2 activity was seen in a 32.5% (164/504) of the PTC studied and this association was statistically significant (P = 0.0244). mTOR signaling complex was also found to be associated with activated AKT and 4E-BP1. In vitro, using Torin2, a second-generation mTOR inhibitor or gene silencing of mTOR expression prevented mTORC1 and mTORC2 activity leading to inactivation of P70S6, 4E-BP1, AKT and Bad. Inhibition of mTOR activity led to downregulation of cyclin D1, a gene regulated by messenger RNA translation via phosphorylation of 4E-BP1. Torin2 treatment also inhibited cell viability and induced caspase-dependent apoptosis via activation of mitochondrial apoptotic pathway in PTC cells. Finally, Torin2 treatment induces anticancer effect on PTC xenograft tumor growth in nude mice via inhibition of mTORC1 and mTORC2 and its associated pathways. Our results suggest that coexpression of mTORC1 and mTORC2 is seen frequently in the clinical PTC samples and dual targeting of mTORC1 and mTORC2 activity may be an attractive therapeutic target for treatment of PTC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1224 Torin2, ≥98% (HPLC)
C24H15F3N4O