EMAIL THIS PAGE TO A FRIEND
Journal of Photochemistry and Photobiology. B, Biology
Malondialdehyde-derived epitopes in human skin result from acute exposure to solar UV and occur in nonmelanoma skin cancer tissue.
PMID 24584085