EMAIL THIS PAGE TO A FRIEND
Testing
DNA aptamers against exon v10 of CD44 inhibit breast cancer cell migration.
PMID 24586375
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB2082
Anti-CD44v10 Antibody, exon v10, clone, Chemicon®, from rabbit