EMAIL THIS PAGE TO A FRIEND
Current eye research
TLR4 contributes to the host response to Klebsiella intraocular infection.
PMID 24588082
Abstract
Klebsiella pneumoniae causes a blinding infection called endogenous endophthalmitis. The role of innate immune recognition of K. pneumoniae in the eye during infection is not known. We hypothesized that intraocular recognition of K. pneumoniae was mediated by Toll-like receptor (TLR)-4 and may be dependent on MagA-regulated hypermucoviscosity. Experimental endophthalmitis was induced in C57BL/6J or TLR4(-/-) mice by intravitreal injection of 100 CFU of wild type or ΔmagA K. pneumoniae. Infection and inflammation were quantified by determining viable K. pneumoniae per eye, retinal responses via electroretinography, myeloperoxidase activity of infiltrating neutrophils and the proinflammatory cytokine and chemokine response. C57BL/6J and TLR4(-/-) mice could not control intraocular wild-type K. pneumoniae growth. TLR4(-/-) mice were less able than C57BL/6J to control the intraocular growth of ΔmagA K. pneumoniae. Retinal function testing suggested that infection with ΔmagA K. pneumoniae resulted in less retinal function loss. There was a TLR4-dependent delay in initial neutrophil recruitment, regardless of the infecting organism. The proinflammatory cytokine/chemokine data supported these results. These findings were not due to an inability of TLR4(-/-) neutrophils to recognize or kill K. pneumoniae. These studies suggest that TLR4 is important in the early intraocular recognition and host response to K. pneumoniae. However, the role of MagA in TLR4-mediated intraocular recognition and subsequent inflammation is less clear.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1240000 Phenylephrine, European Pharmacopoeia (EP) Reference Standard
C9H13NO2