EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
A concomitant loss of dormant origins and FANCC exacerbates genome instability by impairing DNA replication fork progression.
PMID 24589582
Abstract
Accumulating evidence suggests that dormant DNA replication origins play an important role in the recovery of stalled forks. However, their functional interactions with other fork recovery mechanisms have not been tested. We previously reported intrinsic activation of the Fanconi anemia (FA) pathway in a tumor-prone mouse model (Mcm4chaos3) with a 60% loss of dormant origins. To understand this further, we introduced a null allele of Fancc (Fancc-), encoding a member of the FA core complex, into the Mcm4chaos3 background. Primary embryonic fibroblasts double homozygous for Mcm4chaos3 and Fancc- (Mcm4chaos3/chaos3;Fancc-/-) showed significantly increased levels of markers of stalled/collapsed forks compared to either single homozygote. Interestingly, a loss of dormant origins also increased the number of sites in which replication was delayed until prophase, regardless of FA pathway activation. These replication defects coincided with substantially elevated levels of genome instability in Mcm4chaos3/chaos3;Fancc-/- cells, resulting in a high rate of perinatal lethality of Mcm4chaos3/chaos3;Fancc-/- mice and the accelerated tumorigenesis of surviving mice. Together, these findings uncover a specialized role of dormant origins in replication completion while also identifying important functional overlaps between dormant origins and the FA pathway in maintaining fork progression, genome stability, normal development and tumor suppression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA052809
Anti-ADGRE1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA000440
Anti-CD45 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
11333062910 Anti-Digoxigenin, from mouse IgG1κ (clone 1.71.256)
11207750910 Anti-Digoxigenin-Rhodamine, Fab fragments, from sheep
SAB1405756
Anti-EMR1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB1407576
Anti-ERCC6L antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA050492
Anti-ERCC6L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411013
Anti-ERCC6L antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
F0305
Anti-FANCD2 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
HPA039972
Anti-FANCI antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2501430
Anti-Lgals3 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406081
Anti-LGALS3 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003162
Anti-LGALS3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2101338
Anti-LGALS3 antibody produced in rabbit, affinity isolated antibody
SAB1406110
Anti-MCM4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB2701721
Anti-mcm4 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA004873
Anti-MCM4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
GW21465 Anti-PTPRC antibody produced in chicken, affinity isolated antibody, buffered aqueous solution
SAB1406400
Anti-RPA2 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA026309
Anti-RPA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2
HPA026306
Anti-RPA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1
HPA008788
Anti-TP53BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
HPA022133
Anti-TP53BP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
PLA0303
Goat anti-53BP1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
PLA0298
Goat anti-RPA32 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB4700471 Monoclonal Anti-CD45 antibody produced in mouse, clone MEM-28, purified immunoglobulin, buffered aqueous solution
AMAB90518
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0159, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90519
Monoclonal Anti-CD45 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0160, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700587 Monoclonal Anti-CD45 antibody produced in rat, clone EM-05, purified immunoglobulin, buffered aqueous solution
SAB2702173
Monoclonal Anti-FANCD2 antibody produced in mouse