EMAIL THIS PAGE TO A FRIEND
Environmental Science and Pollution Research International
Survey regarding the occurrence of selected organic micropollutants in the groundwaters of overseas departments.
PMID 24595746