EMAIL THIS PAGE TO A FRIEND
Biotechnology and bioengineering
Affinity precipitation of a monoclonal antibody from an industrial harvest feedstock using an ELP-Z stimuli responsive biopolymer.
PMID 24595842
Abstract
In this work, a proof of concept elastin-like polypeptide-Z domain fusion (ELP-Z) based affinity precipitation process is developed for monoclonal antibody (mAb) purification from industrial harvest feeds. Greater than 99% mAb recoveries are obtained during the initial binding step of the process for both pure mAb and the mAb harvest feeds. Great than 90% overall mAb yields are also obtained for the subsequent elution step of the process with no measurable mAb aggregation. The process is shown to result in more than 2 logs of host cell protein (HCP) and more than 4 logs of DNA clearance from the harvest feed. While the overall mAb yield and HCP clearance for the affinity precipitation process was comparable to Protein A chromatography the DNA clearance was clearly superior. Performance is maintained for mAb final elution concentrations up to 20 g/L, demonstrating the ability of the process to both concentrate and purify the mAb. Effective ELP-Z regeneration is also demonstrated using 0.1 M NaOH with no adverse effect on subsequent capture efficiency. Finally, the reusability of the ELP-Z construct and robustness of the process is demonstrated for up to three purification-regeneration cycles with minimal product and impurity carryover and high yields and purity. This work demonstrates that the ELP-Z based precipitation approach can be successfully employed as an affinity capture step for industrial mAbs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1271000
Arginine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C6H14N4O2 · HCl
PHR1454
Arginine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N4O2 · HCl
RDD001
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
CH5N3 · HCl
PHG0006
Guanidine hydrochloride, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
CH5N3 · HCl
G4505
Guanidine hydrochloride, ≥99% (titration), organic base and chaeotropic agent
CH5N3 · HCl
50933
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
CH5N3 · HCl
G3272
Guanidine hydrochloride, for molecular biology, ≥99%
CH5N3 · HCl
G7153
Guanidine hydrochloride, BioXtra, ≥99.5% (titration)
CH5N3 · HCl
G4630
Guanidine hydrochloride, ≥98%
CH5N3 · HCl
50940
Guanidine hydrochloride, ≥99.0% (AT)
CH5N3 · HCl
50950
Guanidine hydrochloride, ≥98%
CH5N3 · HCl
G9284
Guanidine hydrochloride solution, 8 M in H2O (filtered)
CH5N3 · HCl
50937
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
CH5N3 · HCl
08163
L-Arginine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard
C6H14N4O2 · HCl
A5131
L-Arginine monohydrochloride, reagent grade, ≥98% (HPLC), powder
C6H14N4O2 · HCl
A6969
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H14N4O2 · HCl
A4599
L-Arginine monohydrochloride, PharmaGrade, Ajinomoto, EP, JP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production, suitable for cell culture
C6H14N4O2 · HCl
11039
L-Arginine monohydrochloride, BioUltra, ≥99.5% (AT)
C6H14N4O2 · HCl
90538
L-Arginine monohydrochloride, certified reference material, TraceCERT®
C6H14N4O2 · HCl
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
C0909
Sodium citrate tribasic dihydrate, suitable for amino acid analysis, ≥98% (titration), powder
C6H5Na3O7 · 2H2O
C5920
Sodium citrate tribasic dihydrate, insect cell culture tested
C6H5Na3O7 · 2H2O
71404
Sodium citrate tribasic dihydrate, BioUltra, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71405
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
71406
Sodium citrate tribasic dihydrate, purum p.a., ≥99.0% (NT)
C6H5Na3O7 · 2H2O