EMAIL THIS PAGE TO A FRIEND
British Journal of Pharmacology
Dorsal hippocampal NMDA receptor blockade impairs extinction of naloxone-precipitated conditioned place aversion in acute morphine-treated rats by suppressing ERK and CREB phosphorylation in the basolateral amygdala.
PMID 24597568