EMAIL THIS PAGE TO A FRIEND
Josanpu zasshi = The Japanese journal for midwife
[Naming of a child].
PMID 246050
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
245976
2,4,6-Tri-tert-butylpyridine, 99%
C17H29N