Description
Molecular Formula
Add to Cart
SML1210 Almotriptan malate, ≥98% (HPLC)
C17H25N3O2S · C4H6O5
1013512 Almotriptan malate, United States Pharmacopeia (USP) Reference Standard
C17H25N3O2S · C4H6O5