EMAIL THIS PAGE TO A FRIEND
Biochemistry
Imidazole C-2 hydrogen/deuterium exchange reaction at histidine for probing protein structure and function with matrix-assisted laser desorption ionization mass spectrometry.
PMID 24606199