EMAIL THIS PAGE TO A FRIEND
British journal of haematology
Elevated plasma 2-hydroxyglutarate in acute myeloid leukaemia: association with the IDH1 SNP rs11554137 and severe renal impairment.
PMID 24606602