EMAIL THIS PAGE TO A FRIEND
Fertility and sterility
The artificial ovary: any new step is a step forward.
PMID 24613531
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5386 Fibrin from human plasma, insoluble powder