EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Pharmacological properties and clinical efficacy of tetrabenazine (Choreazine(®)Tablets 12.5 mg), a therapeutic agent for non-rhythmic involuntary movement].
PMID 24614638
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T2952
Tetrabenazine, ≥98% (HPLC), solid
C19H27NO3