EMAIL THIS PAGE TO A FRIEND
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association
Clinical significance of RacGAP1 expression at the invasive front of gastric cancer.
PMID 24615626