EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Enantioseparation of new triadimenol antifungal active compounds by electrokinetic chromatography and molecular modeling study of chiral recognition mechanisms.
PMID 24615979