EMAIL THIS PAGE TO A FRIEND
Journal of medical virology
Coxsackievirus B1 reveals strain specific differences in plasmacytoid dendritic cell mediated immunogenicity.
PMID 24616040
Abstract
Enterovirus infections are usually mild but can also cause severe illnesses and play a role in chronic diseases, such as cardiomyopathies and type 1 diabetes. Host response to the invading virus can markedly modulate the course of the infection, and this response varies between individuals due to the polymorphism of immune response genes. However, it is currently not known if virus strains also differ in their ability to stimulate the host immune system. Coxsackievirus B1 (CBV1) causes severe epidemics in young infants and it has recently been connected with type 1 diabetes in seroepidemiological studies. This study evaluated the ability of different field isolates of CBV1 to induce innate immune responses in PBMCs. CBV1 strains differed markedly in their capacity to induce innate immune responses. Out of the 18 tested CBV1 strains two induced exceptionally strong alpha interferon (IFN-α) response in PBMC cultures. The responding cell type was found to be the plasmacytoid dendritic cell. Such a strong innate immune response was accompanied by an up-regulation of several other immune response genes and secretion of cytokines, which modulate inflammation, and adaptive immune responses. These results suggest that enterovirus-induced immune activation depends on the virus strain. It is possible that the immunotype of the virus modulates the course of the infection and plays a role in the pathogenesis of chronic immune-mediated enterovirus diseases.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4300686
Anti-CD3E antibody produced in rabbit, affinity isolated antibody
HPA004252
Anti-CD4 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA004472
Anti-CD4 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1405577
Anti-CD8A antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA037756
Anti-CD8A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
RAB0244
Human IL-10 ELISA Kit, for serum, plasma, cell culture supernatant and urine
RAB0249
Human IL-10 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
C7048
Monoclonal Anti-CD3 antibody produced in mouse, clone UCHT-1, purified immunoglobulin, buffered aqueous solution
SAB4700044 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-57, purified immunoglobulin, buffered aqueous solution
SAB4700045 Monoclonal Anti-CD3 antibody produced in mouse, clone MEM-92, purified immunoglobulin, buffered aqueous solution
SAB4700050 Monoclonal Anti-CD3 antibody produced in mouse, clone APA1/1, purified immunoglobulin, buffered aqueous solution
WH0000916M4
Monoclonal Anti-CD3E antibody produced in mouse, clone 4C1, purified immunoglobulin, buffered aqueous solution
AMAB90876
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1466, purified immunoglobulin, buffered aqueous glycerol solution
AMAB90879
Monoclonal Anti-CD3E antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL1497, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700059
Monoclonal Anti-CD4 , (N-terminal) antibody produced in mouse, clone MEM-241, purified immunoglobulin, buffered aqueous solution
C1805 Monoclonal Anti-CD4 antibody produced in mouse, clone Q4120, purified from hybridoma cell culture
SAB4100489
Monoclonal Anti-CD4 antibody produced in mouse, clone CD4 -A409, culture supernatant
SAB4700054 Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-16, purified immunoglobulin, buffered aqueous solution
SAB4700065
Monoclonal Anti-CD4 antibody produced in mouse, clone MEM-115, purified immunoglobulin, buffered aqueous solution
SAB4700733 Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
AMAB90754
Monoclonal Anti-CD4 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0395, purified immunoglobulin, buffered aqueous glycerol solution
SAB4700449 Monoclonal Anti-CD56 antibody produced in mouse, clone MEM-188, purified immunoglobulin, buffered aqueous solution
SAB4700089 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-87, purified immunoglobulin, buffered aqueous solution
SAB4700084 Monoclonal Anti-CD8 antibody produced in mouse, clone MEM-31, purified immunoglobulin, buffered aqueous solution
C7423
Monoclonal Anti-CD8 antibody produced in mouse, clone UCHT-4, purified immunoglobulin, buffered aqueous solution
SAB1402142
Monoclonal Anti-CD8A antibody produced in mouse, clone 4B9, purified immunoglobulin, buffered aqueous solution
WH0004684M1
Monoclonal Anti-NCAM1 antibody produced in mouse, clone 3G12, purified immunoglobulin, buffered aqueous solution
RAB0245
Mouse IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0246
Rat IL-10 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0247
Rat IL-10 ELISA Kit, for cell and tissue lysates