EMAIL THIS PAGE TO A FRIEND
Cytometry. Part A : the Journal of the International Society For Analytical Cytology
Altered cytological parameters in buccal cells from individuals with mild cognitive impairment and Alzheimer's disease.
PMID 24616437
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
538728
Triethyl phosphate, ReagentPlus®, ≥99.8%
C6H15O4P